Merck
  • Molecular stress relief through a force-induced irreversible extension in polymer contour length.
Molecular stress relief through a force-induced irreversible extension in polymer contour length.
Journal of the American Chemical Society (2010-10-28)
Dong Wu, Jeremy M Lenhardt, Ashley L Black, Boris B Akhremitchev, Stephen L Craig
ABSTRACT
Single-molecule force spectroscopy is used to observe the irreversible extension of a gem-dibromocyclopropane (gDBC)-functionalized polybutadiene under tension, a process akin to polymer necking at a single-molecule level. The extension of close to 28% in the contour length of the polymer backbone occurs at roughly 1.2 nN (tip velocity of 3 μm/s) and is attributed to the force-induced isomerization of the gDBCs into 2,3-dibromoalkenes. The rearrangement represents a possible new mechanism for localized stress relief in polymers and polymer networks under load, and the quantification of the force dependency provides a benchmark value for further studies of mechanically triggered chemistry in bulk polymers.
Sigma-Aldrich
Polybutadiene, average Mn ~5,000
Sigma-Aldrich
Polybutadiene, predominantly 1,2-addition, approx. 90% 1,2-vinyl
Sigma-Aldrich
Polybutadiene, cis, average Mw 200,000-300,000