Merck
  • Addendum to Fragrance material review on Nerolidol (isomer unspecified).
Addendum to Fragrance material review on Nerolidol (isomer unspecified).
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-02-10)
D McGinty, C S Letizia, A M Api
ABSTRACT
An addendum to the toxicologic and dermatologic review of Nerolidol (isomer unspecified) when used as a fragrance ingredient is presented.
Product Description
Supelco
trans-Nerolidol, analytical standard
Sigma-Aldrich
cis-Nerolidol, ≥96.0% (GC)
Sigma-Aldrich
Nerolidol, mixture of cis and trans, ≥97%, stabilized, FG
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans