Merck
  • Primary amino acid lithium salt as a catalyst for asymmetric Michael addition of isobutyraldehyde with beta-nitroalkenes.
Primary amino acid lithium salt as a catalyst for asymmetric Michael addition of isobutyraldehyde with beta-nitroalkenes.
Chemical communications (Cambridge, England) (2008-12-17)
Atsushi Sato, Masanori Yoshida, Shoji Hara
ABSTRACT
Phenylalanine lithium salt was found to be an effective catalyst for asymmetric Michael addition of isobutyraldehyde with beta-nitroalkenes to give quaternary carbon-containing nitroalkanes.
Sigma-Aldrich
Isobutyraldehyde, dry, 98%
Sigma-Aldrich
Isobutyraldehyde, redistilled, ≥99.5%
Sigma-Aldrich
Isobutyraldehyde, 98%
Sigma-Aldrich
Isobutyraldehyde, ≥98%, FG
Sigma-Aldrich
Isobutyraldehyde, natural, 96%, FG
Sigma-Aldrich
Isobutyraldehyde, ≥99%