Merck
  • Solubilisation of drugs in worm-like micelles of block copolymers of ethylene oxide and 1,2-butylene oxide in aqueous solution.
Solubilisation of drugs in worm-like micelles of block copolymers of ethylene oxide and 1,2-butylene oxide in aqueous solution.
International journal of pharmaceutics (2007-12-08)
Zhengyuan Zhou, Chiraphon Chaibundit, Antony D'Emanuele, Kieran Lennon, David Attwood, Colin Booth
ABSTRACT
Ethylene oxide and 1,2-butylene oxide were sequentially polymerised to form the diblock copolymer E13B10 (E=oxyethylene, B=oxybutylene, subscripts denote number-average block lengths in repeat units). Dynamic and static light scattering over the temperature range 10-30 degrees C demonstrated a transition from compact (spheroidal) micelles to larger, more elongated (worm-like) micelles with temperature increase above a critical onset temperature of about 20 degrees C. Determination of the solubilisation capacity for griseofulvin, carbamazepine and spironolactone of dilute micellar solutions of this copolymer, together with those of E11B8 and E17B12 block copolymers (which also show the sphere-to-worm transition), allowed investigation of the influence on solubilisation characteristics of hydrophobic block length and temperature. The extent of solubilisation at 25 degrees C of the poorly water-soluble drug spironolactone increased linearly with increase of hydrophobic block length, attributable to a concomitant increase in the proportion of worm-like micelles in solution.
Sigma-Aldrich
(S)-(−)-1,2-Epoxybutane, 98%
Sigma-Aldrich
1,2-Epoxybutane, ≥99%
Sigma-Aldrich
1,2-Epoxybutane, 99%
Sigma-Aldrich
(R)-(+)-1,2-Epoxybutane, 98%