Merck
  • Strawberry pelargonidin glycosides are excreted in urine as intact glycosides and glucuronidated pelargonidin derivatives in rats.
Strawberry pelargonidin glycosides are excreted in urine as intact glycosides and glucuronidated pelargonidin derivatives in rats.
The British journal of nutrition (2007-06-15)
Catherine Felgines, Odile Texier, Catherine Besson, Bernard Lyan, Jean-Louis Lamaison, Augustin Scalbert
ABSTRACT
Anthocyanins are natural dietary pigments with a wide array of biological properties that are possibly involved in the prevention of various diseases. These properties depend on their absorption and metabolism in the body. In the present study we first examined the gastric and intestinal absorption of pelargonidin 3-glucoside (Pg 3-glc) using rat in situ models. A high proportion of Pg 3-glc was rapidly absorbed from both the stomach (23 %) and small intestine (24 %). Its metabolism was further studied by feeding rats during 8 d with a diet enriched in freeze-dried strawberries. Only low amounts of total anthocyanins were recovered in 24 h urine (0.163 (SEM 0.013) % of ingested anthocyanins; n 8). Strawberry anthocyanins were analysed in urine by HPLC-electrospray ionisation-tandem MS. Similar proportions of intact glycosides (about 53 %) and glucuronidated metabolites (about 47 %) were found. Pg 3-glc was thus glucuronidated to a larger extent than cyanidin 3-glucoside. These results highlight the influence of the aglycone structure on anthocyanin metabolism.
Sigma-Aldrich
Callistephin chloride, ≥95.0% (HPLC)
Sigma-Aldrich
Callistephin chloride, ≥97.0% (HPLC)