- An ultrasensitive method for the determination of thiouracil and phenylthiouracil using capillary zone electrophoresis and laser-induced fluorescence detection.
An ultrasensitive method for the determination of thiouracil and phenylthiouracil using capillary zone electrophoresis and laser-induced fluorescence detection.
Electrophoresis (2005-05-17)
Tomás Pérez-Ruiz, Carmen Martínez-Lozano, Antonio Sanz, Raquel Galera
PMID15895465
ABSTRACT
This paper reports the development of a method based on capillary electrophoresis with laser-induced fluorescence detection for the simultaneous determination of thiouracil (TU) and phenylthiouracil (PhTU) with high sensitivity (nanomolar range, i.e., attomoles detected). After derivatization with 5-iodoacetamidofluorescein, the analytes were separated by capillary zone electrophoresis using 20 mM phosphate buffer (pH 10.0) and quantified by fluorescence detection. The linearity range, precision, recovery, and detection limits were determined, and the method was shown to be applicable for the determination of TU and PhTU in spiked feed samples and urine.