Merck
Volatile organic compounds in the surface waters of northern Greece.
Chemosphere (2000-02-09)
M N Kostopoulou, S K Golfinopoulos, A D Nikolaou, N K Xilourgidis, T D Lekkas
ABSTRACT
An investigation into the occurrence of volatile organic compounds (VOCs) was conducted for a period of two years in the surface waters of Northern Greece. Samples from four rivers and five lakes were taken seasonally and analyzed for VOCs. The analysis has been performed by purge-and-trap (PAT) gas chromatographic-mass spectrometric (GC-MS) technique. The most commonly encountered VOCs in surface waters were chloroform, carbon tetrachloride, trichloroethylene, dichlorobromomethane, tetrachloroethylene, and chlorodibromomethane.
Product Description
Supelco
Dibromochloromethane solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dibromochloromethane, analytical standard
Sigma-Aldrich
Dibromochloromethane, 97%
Supelco
Dibromochloromethane, analytical standard