Merck
  • Utilization of tetrabutylammonium triphenyldifluorosilicate (TBAT) in the synthesis of 6-fluoropurine nucleosides.
Utilization of tetrabutylammonium triphenyldifluorosilicate (TBAT) in the synthesis of 6-fluoropurine nucleosides.
Nucleosides & nucleotides (2000-01-05)
V Gurvich, H Y Kim, R P Hodge, C M Harris, T M Harris
ABSTRACT
Tetrabutylammonium triphenydifluorosilicate (TBAT) has been found to be a useful reagent for the conversion of 6-chloropurine nucleosides to 6-fluoropurine derivatives. The 6-chloropurine nucleosides were reacted with trimethylamine to form quaternary trimethylammonium salts which were treated in situ with TBAT in DMF to effect conversion to the 6-fluoro derivatives in yields of 59-72%.
Sigma-Aldrich
6-Chloropurine, ≥99%
Sigma-Aldrich
6-Chloropurine, 99%