Merck
  • Isoflurane impairs olfaction by increasing neuronal activity in the olfactory bulb.
Isoflurane impairs olfaction by increasing neuronal activity in the olfactory bulb.
Acta physiologica (Oxford, England) (2023-06-18)
Hanyu Fu, Jingwei Zhou, Shan Li, Ying Zhang, Zhiyun Chen, Yingying Yang, Anan Li, Dejuan Wang
ABSTRACT
General anesthesia can induce cognitive deficits in both humans and rodents, correlating with pathological alterations in the hippocampus. However, whether general anesthesia affects olfactory behaviors remains controversial as clinical studies have produced inconsistent results. Therefore, we aimed to investigate how olfactory behaviors and neuronal activity are affected by isoflurane exposure in adult mice. The olfactory detection test, olfactory sensitivity test, and olfactory preference/avoidance test were used to examine olfactory function. In vivo electrophysiology was performed in awake, head-fixed mice to record single-unit spiking and local field potentials in the olfactory bulb (OB). We also performed patch-clamp recordings of mitral cell activity. For morphological studies, immunofluorescence and Golgi-Cox staining were applied. Repeated exposure to isoflurane impaired olfactory detection in adult mice. The main olfactory epithelium, the first region exposed to anesthetics, displayed increased proliferation of basal stem cells. In the OB, a crucial hub for olfactory processing, repeated isoflurane exposure increased the odor responses of mitral/tufted cells. Furthermore, the odor-evoked high gamma response was decreased after isoflurane exposure. Whole-cell recordings further indicated that repeated isoflurane exposure increased the excitability of mitral cells, which may be due to weakened inhibitory input in isoflurane-exposed mice. In addition, elevated astrocyte activation and glutamate transporter-1 expression in the OB were observed in isoflurane-exposed mice. Our findings indicate that repeated isoflurane exposure impairs olfactory detection by increasing neuronal activity in the OB in adult mice.
Sigma-Aldrich
Anti-PSD-95 (Ab-2) Mouse mAb (7E3-1B8), liquid, clone 7E3-1B8, Calbiochem®
Pricing and availability is not currently available.