Merck
  • Extra-virgin olive oil ameliorates high-fat diet-induced seminal and testicular disorders by modulating the cholesterol pathway.
Extra-virgin olive oil ameliorates high-fat diet-induced seminal and testicular disorders by modulating the cholesterol pathway.
Andrology (2023-01-26)
Abi Karenina Funes, María Virginia Avena, Jorge Ibañez, Layla Simón, Leonor Ituarte, Regina Colombo, Adrián Roldán, María Inés Conte, María Ángeles Monclus, Paola Boarelli, Miguel Walter Fornés, Tania Estefanía Saez Lancellotti
ABSTRACT
Rabbits are sensitive to dietary cholesterol and rapidly develop hypercholesterolemia, leading to prominent subfertility. Sterol regulatory element-binding protein isoform 2 drives the intracellular cholesterol pathway in many tissues, including the testicles. Its abnormal regulation could be the mainly responsible for the failure of suppressing cholesterol synthesis in a cholesterol-enriched environment, ultimately leading to testicular and seminal alterations. However, extra-virgin olive oil consumption has beneficial properties that promote lowering of cholesterol levels, including the recovery of seminal parameters altered under a high-fat diet. Our goal was to investigate the effects of high-fat diet supplementation with extra-virgin olive oil at the molecular level on rabbit testes, by analyzing sterol regulatory element-binding protein isoform 2 protein and its corresponding downstream effectors. During 12 months, male rabbits were fed a control diet, high-fat diet, or 6-month high-fat diet followed by 6-month high-fat diet plus extra-virgin olive oil. Serum lipids, testosterone levels, bodyweight, and seminal parameters were tested. The mRNA and protein levels of sterol regulatory element-binding protein isoform 2, 3-hydroxy-3-methyl-glutaryl-coenzyme A reductase, and low-density lipoprotein receptor were determined by semi-quantitative polymerase chain reaction and Western blotting techniques. The expression pattern of sterol regulatory element-binding protein isoform 2 protein in the rabbit testicles was studied by indirect immunofluorescence. In addition, testicular cholesterol was detected and quantified by filipin staining and gas chromatography. The data showed that the addition of extra-virgin olive oil to high-fat diet reduced testicular cholesterol levels and recovered the expression of sterol regulatory element-binding protein isoform 2, 3-hydroxy-3-methyl-glutaryl-coenzyme A reductase, and low-density lipoprotein receptor initially altered by the high-fat diet. The combination of high-fat diet with extra-virgin olive oil encourages testicular recovery by modifying the expression of the enzymes related to intracellular cholesterol management.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution