Merck
Delivery of Active AKT1 to Human Cells.
Cells (2022-12-12)
Tarana Siddika, Nileeka Balasuriya, Mallory I Frederick, Peter Rozik, Ilka U Heinemann, Patrick O'Donoghue
ABSTRACT
Protein kinase B (AKT1) is a serine/threonine kinase and central transducer of cell survival pathways. Typical approaches to study AKT1 biology in cells rely on growth factor or insulin stimulation that activates AKT1 via phosphorylation at two key regulatory sites (Thr308, Ser473), yet cell stimulation also activates many other kinases. To produce cells with specific AKT1 activity, we developed a novel system to deliver active AKT1 to human cells. We recently established a method to produce AKT1 phospho-variants from Escherichia coli with programmed phosphorylation. Here, we fused AKT1 with an N-terminal cell penetrating peptide tag derived from the human immunodeficiency virus trans-activator of transcription (TAT) protein. The TAT-tag did not alter AKT1 kinase activity and was necessary and sufficient to rapidly deliver AKT1 protein variants that persisted in human cells for 24 h without the need to use transfection reagents. TAT-pAKT1T308 induced selective phosphorylation of the known AKT1 substrate GSK-3α, but not GSK-3β, and downstream stimulation of the AKT1 pathway as evidenced by phosphorylation of ribosomal protein S6 at Ser240/244. The data demonstrate efficient delivery of AKT1 with programmed phosphorylation to human cells, thus establishing a cell-based model system to investigate signaling that is dependent on AKT1 activity.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Fc, Chemicon®, from goat
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey