Merck
  • LRG1 Expression Is Elevated in the Eyes of Patients with Neovascular Age-Related Macular Degeneration.
LRG1 Expression Is Elevated in the Eyes of Patients with Neovascular Age-Related Macular Degeneration.
International journal of molecular sciences (2021-08-28)
Lucia Mundo, Gian Marco Tosi, Stefano Lazzi, Grazia Pertile, Barbara Parolini, Giovanni Neri, Matteo Posarelli, Elena De Benedetto, Tommaso Bacci, Ennio Silvestri, Maria Chiara Siciliano, Stefano Barbera, Maurizio Orlandini, John Greenwood, Stephen E Moss, Federico Galvagni
ABSTRACT
Leucine-rich a-2-glycoprotein 1 (LRG1) is a candidate therapeutic target for treating the neovascular form of age-related macular degeneration (nvAMD). In this study we examined the expression of LRG1 in eyes of nvAMD patients. Choroidal neovascular membranes (CNVMs) from patients who underwent submacular surgery for retinal pigment epithelium-choroid graft transplantation were collected from 5 nvAMD patients without any prior intravitreal anti-VEGF injection, and from six patients who received intravitreal anti-VEGF injections before surgery. As controls free of nvAMD, retina sections were obtained from the eyes resected from a patient with lacrimal sac tumor and from a patient with neuroblastoma. CNVMs were immunostained for CD34, LRG1, and α-smooth muscle actin (α-SMA). Aqueous humor samples were collected from 58 untreated-naïve nvAMD patients prior to the intravitreal injection of anti-VEGF and 51 age-matched cataract control patients, and LRG1 concentration was measured by ELISA. The level of LRG1 immunostaining is frequently high in both the endothelial cells of the blood vessels, and myofibroblasts in the surrounding tissue of CNVMs of treatment-naïve nvAMD patients. Furthermore, the average concentration of LRG1 was significantly higher in the aqueous humor of nvAMD patients than in controls. These observations provide a strong experimental basis and scientific rationale for the progression of a therapeutic anti-LRG1 monoclonal antibody into clinical trials with patients with nvAMD.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Biotin antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
EN300-7447613-50MG50 mg
Ships in 14 days - from Aldrich Partner.
CZK 9,607.50
EN300-7447613-100MG100 mg
Ships in 14 days - from Aldrich Partner.
CZK 13,702.50
EN300-7447613-250MG250 mg
Ships in 14 days - from Aldrich Partner.
CZK 19,032.50
EN300-7447613-500MG500 mg
Ships in 14 days - from Aldrich Partner.
CZK 29,400.00
EN300-7447613-1G1 g
Ships in 14 days - from Aldrich Partner.
CZK 37,362.50
EN300-7447613-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
CZK 72,170.00
To order products, please contact your local dealer.
Sigma-Aldrich
Anti-LRG1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
EN300-7447613-50MG50 mg
Ships in 14 days - from Aldrich Partner.
CZK 9,607.50
EN300-7447613-100MG100 mg
Ships in 14 days - from Aldrich Partner.
CZK 13,702.50
EN300-7447613-250MG250 mg
Ships in 14 days - from Aldrich Partner.
CZK 19,032.50
EN300-7447613-500MG500 mg
Ships in 14 days - from Aldrich Partner.
CZK 29,400.00
EN300-7447613-1G1 g
Ships in 14 days - from Aldrich Partner.
CZK 37,362.50
EN300-7447613-2.5G2.5 g
Ships in 14 days - from Aldrich Partner.
CZK 72,170.00
To order products, please contact your local dealer.