Merck
  • Toward reproducible, scalable, and robust data analysis across multiplex tissue imaging platforms.
Toward reproducible, scalable, and robust data analysis across multiplex tissue imaging platforms.
Cell reports methods (2021-09-07)
Erik A Burlingame, Jennifer Eng, Guillaume Thibault, Koei Chin, Joe W Gray, Young Hwan Chang
ABSTRACT
The emergence of megascale single-cell multiplex tissue imaging (MTI) datasets necessitates reproducible, scalable, and robust tools for cell phenotyping and spatial analysis. We developed open-source, graphics processing unit (GPU)-accelerated tools for intensity normalization, phenotyping, and microenvironment characterization. We deploy the toolkit on a human breast cancer (BC) tissue microarray stained by cyclic immunofluorescence and present the first cross-validation of breast cancer cell phenotypes derived by using two different MTI platforms. Finally, we demonstrate an integrative phenotypic and spatial analysis revealing BC subtype-specific features.
Sigma-Aldrich
Monoclonal Anti-Lamin A/C antibody produced in mouse, clone 4C11, purified from hybridoma cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Anti-Androgen Receptor Antibody, Alexa Fluor 555 Conjugate, from rabbit, ALEXA FLUOR 555