Merck
Discovery and characterization of an endogenous CXCR4 antagonist.
Cell reports (2015-04-30)
Onofrio Zirafi, Kyeong-Ae Kim, Ludger Ständker, Katharina B Mohr, Daniel Sauter, Anke Heigele, Silvia F Kluge, Eliza Wiercinska, Doreen Chudziak, Rudolf Richter, Barbara Moepps, Peter Gierschik, Virag Vas, Hartmut Geiger, Markus Lamla, Tanja Weil, Timo Burster, Andreas Zgraja, Francois Daubeuf, Nelly Frossard, Muriel Hachet-Haas, Fabian Heunisch, Christoph Reichetzeder, Jean-Luc Galzi, Javier Pérez-Castells, Angeles Canales-Mayordomo, Jesus Jiménez-Barbero, Guillermo Giménez-Gallego, Marion Schneider, James Shorter, Amalio Telenti, Berthold Hocher, Wolf-Georg Forssmann, Halvard Bonig, Frank Kirchhoff, Jan Münch
ABSTRACT
CXCL12-CXCR4 signaling controls multiple physiological processes and its dysregulation is associated with cancers and inflammatory diseases. To discover as-yet-unknown endogenous ligands of CXCR4, we screened a blood-derived peptide library for inhibitors of CXCR4-tropic HIV-1 strains. This approach identified a 16 amino acid fragment of serum albumin as an effective and highly specific CXCR4 antagonist. The endogenous peptide, termed EPI-X4, is evolutionarily conserved and generated from the highly abundant albumin precursor by pH-regulated proteases. EPI-X4 forms an unusual lasso-like structure and antagonizes CXCL12-induced tumor cell migration, mobilizes stem cells, and suppresses inflammatory responses in mice. Furthermore, the peptide is abundant in the urine of patients with inflammatory kidney diseases and may serve as a biomarker. Our results identify EPI-X4 as a key regulator of CXCR4 signaling and introduce proteolysis of an abundant precursor protein as an alternative concept for chemokine receptor regulation.
Sigma-Aldrich
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97.0% (HPLC)
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
Bestatin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Albumin, Human Serum, Non-denatured
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution