Merck
Synthesis and antimicrobial evaluation of new benzofuran derivatives.
European journal of medicinal chemistry (2011-05-17)
Xizhen Jiang, Wenlu Liu, Wei Zhang, Faqin Jiang, Zhe Gao, Hao Zhuang, Lei Fu
ABSTRACT
Thirteen compounds, based on benzofuran skeleton bearing aryl substituents at its C-3 position through methanone linker, were synthesized and screened for their antibacterial and antifungal activities against four bacteria Escherichia coli, Staphylococcus aureus, Methicillin-resistant S.aureus, Bacillus subtilis, and a fungus Candida albicans. Four hydrophobic benzofuran analogs were found to exhibit favorable antibacterial activities (MIC(80) = 0.39-3.12 μg/mL), which were better than the control drugs.
Sigma-Aldrich
Penicillin G sodium salt, ~1650 U/mg
Sigma-Aldrich
Penicillin G sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(±)-Miconazole nitrate salt, imidazole antibiotic
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)