Merck
  • Innovative Ultrasound-Assisted Approaches towards Reduction of Heavy Metals and Iodine in Macroalgal Biomass.
Innovative Ultrasound-Assisted Approaches towards Reduction of Heavy Metals and Iodine in Macroalgal Biomass.
Foods (Basel, Switzerland) (2021-04-04)
Estefanía Noriega-Fernández, Izumi Sone, Leire Astráin-Redín, Leena Prabhu, Morten Sivertsvik, Ignacio Álvarez, Guillermo Cebrián
ABSTRACT
The aim of this work was to evaluate the potential of ultrasound (US), alone or in combination with mild heating and/or EDTA towards reduction of As, Cd, I, and Hg content of Laminaria hyperborea. Concentrations of As, Cd, I, and Hg of 56.29, 0.596, 7340, and <0.01 mg kg-1 of dry weight, respectively, were found in L. hyperborea blades. Treatment with US at 50 °C increased approx. 2-fold the amount of As released, although did not affect significantly the content of Cd or I, as compared to control (no US) samples. Reducing the temperature to 8 °C significantly decreased the effect of US, but heating at 80 °C did not cause a significant effect as compared to treatments at 50 °C. On the other hand, treatment with 0.1 N EDTA at 50 °C enhanced the percentage of Cd released by approximately 7-fold, regardless of sonication. In the present work, the combination of US and EDTA at 50 °C for 5 min led to a significant reduction of the As (32%), Cd (52%) and I (31%) content in L. hyperborea, thus improving the product's safety for consumers.
Sigma-Aldrich
Trimethylphenylammonium hydroxide solution, ~25% in H2O (1.68 M)