Merck
  • Fungal chitin-glucan nanopapers with heavy metal adsorption properties for ultrafiltration of organic solvents and water.
Fungal chitin-glucan nanopapers with heavy metal adsorption properties for ultrafiltration of organic solvents and water.
Carbohydrate polymers (2020-12-07)
Neptun Yousefi, Mitchell Jones, Alexander Bismarck, Andreas Mautner
ABSTRACT
Membranes and filters are essential devices, both in the laboratory for separation of media, solvent recovery, organic solvent and water filtration purposes, and in industrial scale applications, such as the removal of industrial pollutants, e.g. heavy metal ions, from water. Due to their solvent stability, biologically sourced and renewable membrane or filter materials, such as cellulose or chitin, provide a low-cost, sustainable alternative to synthetic materials for organic solvent filtration and water treatment. Here, we investigated the potential of fungal chitin nanopapers derived from A. bisporus (common white-button mushrooms) as ultrafiltration membranes for organic solvents and aqueous solutions and hybrid chitin-cellulose microfibril papers as high permeance adsorptive filters. Fungal chitin constitutes a renewable, easily isolated, and abundant alternative to crustacean chitin. It can be fashioned into solvent stable nanopapers with pore sizes of 10-12 nm, as determined by molecular weight cut-off and rejection of gold nanoparticles, that exhibit high organic solvent permeance, making them a valuable material for organic solvent filtration applications. Addition of cellulose fibres to produce chitin-cellulose hybrid papers extended membrane functionality to water treatment applications, with considerable static and dynamic copper ion adsorption capacities and high permeances that outperformed other biologically derived membranes, while being simpler to produce, naturally porous, and not requiring crosslinking. The simple nanopaper production process coupled with the remarkable filtration properties of the papers for both organic solvent filtration and water treatment applications designates them an environmentally benign alternative to traditional membrane and filter materials.
Sigma-Aldrich
D-(+)-Glucosamine hydrochloride, ≥99% (HPLC), powder
Sigma-Aldrich
Chitin from shrimp shells, practical grade, coarse flakes
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG