Merck
  • New Methodology for the Identification of Metabolites of Saccharides and Cyclitols by Off-Line EC-MALDI-TOF-MS.
New Methodology for the Identification of Metabolites of Saccharides and Cyclitols by Off-Line EC-MALDI-TOF-MS.
International journal of molecular sciences (2020-07-30)
Gulyaim Sagandykova, Justyna Walczak-Skierska, Fernanda Monedeiro, Paweł Pomastowski, Bogusław Buszewski
ABSTRACT
A combination of electrochemistry (EC) and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (off-line EC-MALDI-TOF-MS) was applied for determination of the studied biologically active compounds (D-glucose, D-fructose, D-galactose, D-pinitol, L-chiro-inositol, and myo-inositol) and their possible electrochemical metabolites. In this work, boron-doped diamond electrode (BDD) was used as a working electrode. MALDI-TOF-MS experiments were carried out (both in positive and negative ion modes and using two matrices) to identify the structures of electrochemical products. This was one of the first applications of the EC system for the generation of electrochemical products produced from saccharides and cyclitols. Moreover, exploratory data analysis approaches (correlation networks, hierarchical cluster analysis, weighted plots) were used in order to present differences/similarities between the obtained spectra, regarding the class of analyzed compounds, ionization modes, and used matrices. This work presents the investigation and comparison of fragmentation patterns of sugars, cyclitols, and their respective products generated through the electrochemistry (EC) process.
Sigma-Aldrich
Cesium triiodide, ≥99.9%
Sigma-Aldrich
D-Pinitol, 95%
Sigma-Aldrich
1-Naphthalenyl hydrazine hydrochloride, AldrichCPR