Merck
  • Identification of HMGA2 inhibitors by AlphaScreen-based ultra-high-throughput screening assays.
Identification of HMGA2 inhibitors by AlphaScreen-based ultra-high-throughput screening assays.
Scientific reports (2020-11-04)
Linjia Su, Nadezda Bryan, Sabrina Battista, Juliano Freitas, Alyssa Garabedian, Federica D'Alessio, Miriam Romano, Fabiana Falanga, Alfredo Fusco, Lidia Kos, Jeremy Chambers, Francisco Fernandez-Lima, Prem P Chapagain, Stefan Vasile, Layton Smith, Fenfei Leng
ABSTRACT
The mammalian high mobility group protein AT-hook 2 (HMGA2) is a multi-functional DNA-binding protein that plays important roles in tumorigenesis and adipogenesis. Previous results showed that HMGA2 is a potential therapeutic target of anticancer and anti-obesity drugs by inhibiting its DNA-binding activities. Here we report the development of a miniaturized, automated AlphaScreen ultra-high-throughput screening assay to identify inhibitors targeting HMGA2-DNA interactions. After screening the LOPAC1280 compound library, we identified several compounds that strongly inhibit HMGA2-DNA interactions including suramin, a century-old, negatively charged antiparasitic drug. Our results show that the inhibition is likely through suramin binding to the "AT-hook" DNA-binding motifs and therefore preventing HMGA2 from binding to the minor groove of AT-rich DNA sequences. Since HMGA1 proteins also carry multiple "AT-hook" DNA-binding motifs, suramin is expected to inhibit HMGA1-DNA interactions as well. Biochemical and biophysical studies show that charge-charge interactions and hydrogen bonding between the suramin sulfonated groups and Arg/Lys residues play critical roles in the binding of suramin to the "AT-hook" DNA-binding motifs. Furthermore, our results suggest that HMGA2 may be one of suramin's cellular targets.
Sigma-Aldrich
CHAPS hydrate, ≥98% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
LOPAC®1280