Merck
BFRF1 protein is involved in EBV-mediated autophagy manipulation.
Microbes and infection (2020-09-04)
Roberta Gonnella, Marzia Dimarco, Giuseppina A Farina, Roberta Santarelli, Sandro Valia, Alberto Faggioni, Antonio Angeloni, Mara Cirone, Antonella Farina
ABSTRACT
Viral egress and autophagy are two mechanisms that seem to be strictly connected in Herpesviruses's biology. Several data suggest that the autophagic machinery facilitates the egress of viral capsids and thus the production of new infectious particles. In the Herpesvirus family, viral nuclear egress is controlled and organized by a well conserved group of proteins named Nuclear Egress Complex (NEC). In the case of EBV, NEC is composed by BFRF1 and BFLF2 proteins, although the alterations of the nuclear host cell architecture are mainly driven by BFRF1, a multifunctional viral protein anchored to the inner nuclear membrane of the host cell. BFRF1 shares a peculiar distribution with several nuclear components and with them it strictly interacts. In this study, we investigated the possible role of BFRF1 in manipulating autophagy, pathway that possibly originates from nucleus, regulating the interplay between autophagy and viral egress.
Product Description
Millipore
BCA Protein Assay Kit, The BCA protein assay is a simple and reliable protein quantification method.
Pricing and availability is not currently available.