Merck
  • SB 203580 is a specific inhibitor of a MAP kinase homologue which is stimulated by cellular stresses and interleukin-1.
SB 203580 is a specific inhibitor of a MAP kinase homologue which is stimulated by cellular stresses and interleukin-1.
FEBS letters (1995-05-08)
A Cuenda, J Rouse, Y N Doza, R Meier, P Cohen, T F Gallagher, P R Young, J C Lee
ABSTRACT
A class of pyridinyl imidazoles inhibit the MAP kinase homologue, termed here reactivating kinase (RK) [Lee et al. (1994) Nature 372, 739-746]. We now show that one of these compounds (SB 203580) inhibits RK in vitro (IC50 = 0.6 microM), suppresses the activation of MAPKAP kinase-2 and prevents the phosphorylation of heat shock protein (HSP) 27 in response to interleukin-1, cellular stresses and bacterial endotoxin in vivo. These results establish that MAPKAP kinase-2 is a physiological RK substrate, and that HSP27 is phosphorylated by MAPKAP kinase-2 in vivo. The specificity of SB 203580 was indicated by its failure to inhibit 12 other protein kinases in vitro, and by its lack of effect on the activation of RK kinase and other MAP kinase cascades in vivo. We suggest that SB 203580 will be useful for identifying other physiological roles and targets of RK and MAPKAP kinase-2.
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
p38/SAPK2 Inhibitor (SB 203580), The p38/SAPK2 Inhibitor (SB 203580) controls the biological activity of p38/SAPK2. This small molecule/inhibitor is primarily used for Biochemicals applications.
Sigma-Aldrich
p38α/SAPK2a Protein, active, 10 µg, Active, N-terminal GST tagged, recombinant, full length p38α/SAPK2a, for use in Kinase Assays.
Sigma-Aldrich
p38β2/SAPK2b2 Protein, active, 10 µg, Active, N-terminal GST fusion protein corresponding to full length human p38β2/SAPK2b2 activated with MKK6. For use in Kinase Assays.