Merck
  • (IPr)Pd(acac)Cl: an easily synthesized, efficient, and versatile precatalyst for C-N and C-C bond formation.
(IPr)Pd(acac)Cl: an easily synthesized, efficient, and versatile precatalyst for C-N and C-C bond formation.
The Journal of organic chemistry (2006-05-06)
Nicolas Marion, Elise C Ecarnot, Oscar Navarro, Dino Amoroso, Andrew Bell, Steven P Nolan
ABSTRACT
A very straightforward synthesis of (IPr)Pd(acac)Cl from two commercially available starting materials, Pd(acac)2 and IPr.HCl [acac = acetylacetonate; IPr = N,N'-bis(2,6-diisopropylphenyl)imidazol-2-ylidene], has been developed. The resulting complex, (IPr)Pd(acac)Cl (1), has proven to be a highly active PdII precatalyst in the Buchwald-Hartwig and the alpha-ketone arylation reactions. A wide range of substrates has been screened, including unactivated, sterically hindered, and heterocyclic aryl chlorides.
Sigma-Aldrich
Pd(IPr)(acac)Cl
Sigma-Aldrich
Palladium(II) acetylacetonate, ≥99.9% trace metals basis