Merck
  • An HIV-Tat inducible mouse model system of childhood HIV-associated nephropathy.
An HIV-Tat inducible mouse model system of childhood HIV-associated nephropathy.
Disease models & mechanisms (2020-09-13)
Pingtao Tang, Jharna R Das, Jinliang Li, Jing Yu, Patricio E Ray
ABSTRACT
Modern antiretroviral therapies (ART) have decreased the prevalence of HIV-associated nephropathy (HIVAN). Nonetheless, we continue to see children and adolescents with HIVAN all over the world. Furthermore, once HIVAN is established in children, it is difficult to revert its long-term progression, and we need better animal models of childhood HIVAN to test new treatments. To define whether the HIV-1 trans-activator (Tat) gene precipitates HIVAN in young mice, and to develop an inducible mouse model of childhood HIVAN, an HIV-Tat gene cloned from a child with HIVAN was used to generate recombinant adenoviral vectors (rAd-Tat). rAd-Tat and LacZ control vectors (2×109) were expressed in the kidney of newborn wild-type and HIV-transgenic (Tg26) FVB/N mice without significant proteinuria (n=5; 8 per group). Mice were sacrificed 7 and 35 days later to assess their renal outcome, the expression of HIV-genes and growth factors, and markers of cell growth and differentiation by RT-qPCR, immunohistochemistry and/or western blots. HIV-Tat induced the expression of HIV-1 genes and heparin-binding growth factors in the kidney of HIV-Tg26 mice, and precipitated HIVAN in the first month of life. No significant renal changes were detected in wild-type mice infected with rAd-Tat vectors, suggesting that HIV-Tat alone does not induce renal disease. This new mouse model of childhood HIVAN highlights the critical role that HIV-Tat plays in the pathogenesis of HIVAN, and could be used to study the pathogenesis and treatment of HIVAN in children and adolescents.
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Sigma-Aldrich
ApopTag Plus Peroxidase In Situ Apoptosis Kit, The ApopTag Plus Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells by labeling & detecting DNA strand breaks by the indirect TUNEL method.
Sigma-Aldrich
Anti-Wilms′ Tumor Antibody, NT, clone 6F-H2, clone 6F-H2, Chemicon®, from mouse