Merck
  • Mesenchymal Stem and Stromal Cells Harness Macrophage-Derived Amphiregulin to Maintain Tissue Homeostasis.
Mesenchymal Stem and Stromal Cells Harness Macrophage-Derived Amphiregulin to Maintain Tissue Homeostasis.
Cell reports (2020-03-19)
Jung Hwa Ko, Hyeon Ji Kim, Hyun Jeong Jeong, Hyun Ju Lee, Joo Youn Oh
ABSTRACT
The cross-talk between mesenchymal stem and stromal cells (MSCs) and macrophages is critical for the restoration of tissue homeostasis after injury. Here, we demonstrate a pathway through which MSCs instruct macrophages to resolve inflammation and preserve tissue-specific stem cells, leading to homeostasis in mice with autoimmune uveoretinitis and sterile-injury-induced corneal epithelial stem cell deficiency. Distinct from their conventional role in macrophage reprogramming to anti-inflammatory phenotype by a PGE2-dependent mechanism, MSCs enhance the phagocytic activity of macrophages, which partly depends on the uptake of MSC mitochondria-containing extracellular vesicles. The MSC-primed macrophages increase the secretion of amphiregulin (AREG) in a phagocytosis-dependent manner. AREG is essential for MSC-primed macrophages to suppress immune responses through regulatory T (Treg) cells and to protect corneal epithelial stem cells via apoptosis inhibition and proliferation promotion. Hence, the data reveal that MSCs harness macrophage-derived AREG to maintain tissue homeostasis after injury and provide a therapeutic target in immune-mediated disease and regenerative medicine.
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.
Sigma-Aldrich
GW4869, ≥90% (NMR)
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, Rhodamine conjugate, Chemicon®, from goat