Merck
  • Biological characterization of new inhibitors of microsomal PGE synthase-1 in preclinical models of inflammation and vascular tone.
Biological characterization of new inhibitors of microsomal PGE synthase-1 in preclinical models of inflammation and vascular tone.
British journal of pharmacology (2019-08-14)
Karin Larsson, Julia Steinmetz, Filip Bergqvist, Samsul Arefin, Linda Spahiu, Johan Wannberg, Sven-Christian Pawelzik, Ralf Morgenstern, Patric Stenberg, Karolina Kublickiene, Marina Korotkova, Per-Johan Jakobsson
ABSTRACT
Microsomal PGE synthase-1 (mPGES-1), the inducible synthase that catalyses the terminal step in PGE2 biosynthesis, is of high interest as therapeutic target to treat inflammation. Inhibition of mPGES-1 is suggested to be safer than traditional NSAIDs, and recent data demonstrate anti-constrictive effects on vascular tone, indicating new therapeutic opportunities. However, there is a lack of potent mPGES-1 inhibitors lacking interspecies differences for conducting in vivo studies in relevant preclinical disease models. Potency was determined based on the reduction of PGE2 formation in recombinant enzyme assays, cellular assay, human whole blood assay, and air pouch mouse model. Anti-inflammatory properties were assessed by acute paw swelling in a paw oedema rat model. Effect on vascular tone was determined with human ex vivo wire myography. We report five new mPGES-1 inhibitors (named 934, 117, 118, 322, and 323) that selectively inhibit recombinant human and rat mPGES-1 with IC50 values of 10-29 and 67-250 nM respectively. The compounds inhibited PGE2 production in a cellular assay (IC50 values 0.15-0.82 μM) and in a human whole blood assay (IC50 values 3.3-8.7 μM). Moreover, the compounds blocked PGE2 formation in an air pouch mouse model and reduced acute paw swelling in a paw oedema rat model. Human ex vivo wire myography analysis showed reduced adrenergic vasoconstriction after incubation with the compounds. These mPGES-1 inhibitors can be used as refined tools in further investigations of the role of mPGES-1 in inflammation and microvascular disease.
Sigma-Aldrich
NS-398, ≥98% (HPLC), solid