Merck
  • Skin surface lipid composition in women: increased 2,3-oxidosqualene correlates with older age.
Skin surface lipid composition in women: increased 2,3-oxidosqualene correlates with older age.
European journal of dermatology : EJD (2020-04-14)
Aline Rigal, Rime Michael-Jubeli, Armelle Bigouret, Alex Nkengne, Dominique Bertrand, Arlette Baillet-Guffroy, Ali Tfayli
ABSTRACT
The importance of the hydrolipidic film of skin has been well documented, however, few data are available in cases of very old age. Our aim was to characterize the difference in skin surface lipid (SSL) composition between individuals of different age groups. Data were collected from the forehead of 22 young volunteers (18-24 years old) and 18 senior volunteers (70-75 years old). The amount of sebum was obtained by sebumetry. To acquire relevant information about the molecular composition of high complex mixtures, SSLs were analysed in a single run to ensure that the lipid structures remain intact, using high-temperature gas chromatography coupled with mass spectrometry. The major features associated with aged skin were documented. In aged skin, a lower sebum content was observed, together with modification of the relative SSL composition involving a significant reduction in the intensity of many components of the hydrolipidic film. In contrast, the intensity of 2,3-oxidosqualene was shown to increase with an inverse relationship between triglycerides and their hydrolytic products. These adaptations could be related to modifications of enzymatic activity.
Sigma-Aldrich
1-Stearoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Dipalmitin, ≥99.0%
Sigma-Aldrich
Dodecyl arachidate, ≥99%