Merck
  • Exosomal miR-25-3p from mesenchymal stem cells alleviates myocardial infarction by targeting pro-apoptotic proteins and EZH2.
Exosomal miR-25-3p from mesenchymal stem cells alleviates myocardial infarction by targeting pro-apoptotic proteins and EZH2.
Cell death & disease (2020-05-07)
Yi Peng, Ji-Ling Zhao, Zhi-Yong Peng, Wei-Fang Xu, Guo-Long Yu
ABSTRACT
Mesenchymal stem cell (MSC) therapy is a promising approach against myocardial infarction (MI). Studies have demonstrated that MSCs can communicate with other cells by secreting exosomes. In the present study, we aimed to identify exosomal microRNAs that might contribute to MSC-mediated cardioprotective effects. Primary cardiomyocytes were deprived of oxygen and glucose to mimic MI in vitro. For the animal model of MI, the left anterior descending artery was ligated for 1 h, followed by reperfusion for 12 h. MSC-derived exosomes were used to treat primary cardiomyocytes or mice. Cardioprotection-related microRNAs were determined, followed by target gene identification and functional studies with quantitative PCR, western blotting, MTT assay, flow cytometry assay, chromatin immunoprecipitation and dual-luciferase assay. We found that MSC co-culture reduced OGD-induced cardiomyocyte apoptosis and inflammatory responses. Cardioprotection was also observed upon treatment with MSC-derived exosomes in vitro and in vivo. In line with this, exosome uptake led to a significant increase in miR-25-3p in cardiomyocytes. Depletion of miR-25-3p in MSCs abolished the protective effects of exosomes. Mechanistically, miR-25-3p directly targeted the pro-apoptotic genes FASL and PTEN and reduced their protein levels. Moreover, miR-25-3p decreased the levels of EZH2 and H3K27me3, leading to derepression of the cardioprotective gene eNOS as well as the anti-inflammatory gene SOCS3. Inhibition of EZH2 or overexpression of miR-25-3p in cardiomyocytes was sufficient to confer cardioprotective effects in vitro and in vivo. We concluded that exosomal miR-25-3p from MSCs alleviated MI by targeting pro-apoptotic proteins and EZH2.
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
MISSION® esiRNA, targeting human SOCS3
Sigma-Aldrich
Anti-Fas Ligand Antibody, Chemicon®, from rabbit