Merck
  • A bicyclic pentapeptide-based highly potent and selective pan-SIRT1/2/3 inhibitor harboring Nε-thioacetyl-lysine.
A bicyclic pentapeptide-based highly potent and selective pan-SIRT1/2/3 inhibitor harboring Nε-thioacetyl-lysine.
Bioorganic & medicinal chemistry (2020-02-19)
Renwu Li, Lingling Yan, Xun Sun, Weiping Zheng
ABSTRACT
Past few years have seen an active pursuit of the inhibitors for the deacylation catalyzed by the seven human sirtuins (i.e. SIRT1-7) as valuable chemical biological/pharmacological probes of this enzymatic deacylation and lead compounds for developing novel therapeutics for human diseases. In the current study, we prepared eight monocyclic and one bicyclic analogs of a linear pentapeptide-based potent (sub-μM IC50's) pan-SIRT1/2/3 inhibitor Zheng laboratory discovered recently that harbors the catalytic mechanism-based SIRT1/2/3 inhibitory warhead Nε-thioacetyl-lysine at its central position. We found that the bicyclic analog exhibited largely comparable SIRT1/2/3 inhibitory potencies to those of the parent linear pentapeptide, however, the former is proteolytically much more stable than the latter. Moreover, the bicyclic analog displayed very weak inhibition against SIRT5/6/7, was cell permeable, and exhibited an anti-proliferative effect on the human SK-MEL-2 melanoma cells. This bicyclic analog could be a lead for the future development of more potent and still selective pan-SIRT1/2/3 inhibitors whose use in studies on human sirtuin biology, pharmacology, and medicinal chemistry could complement with the use of the potent inhibitors selective for a single human sirtuin.
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
Ethyl dithioacetate, 98%
Sigma-Aldrich
4-Bromobutyric acid, 98%