Merck
  • DLC1 is a direct target of activated YAP/TAZ that drives collective migration and sprouting angiogenesis.
DLC1 is a direct target of activated YAP/TAZ that drives collective migration and sprouting angiogenesis.
Journal of cell science (2020-01-23)
Miesje van der Stoel, Lilian Schimmel, Kalim Nawaz, Anne-Marieke van Stalborch, Annett de Haan, Alexandra Klaus-Bergmann, Erik T Valent, Duco S Koenis, Geerten P van Nieuw Amerongen, Carlie J de Vries, Vivian de Waard, Martijn Gloerich, Jaap D van Buul, Stephan Huveneers
ABSTRACT
Endothelial YAP/TAZ (YAP is also known as YAP1, and TAZ as WWTR1) signaling is crucial for sprouting angiogenesis and vascular homeostasis. However, the underlying molecular mechanisms that explain how YAP/TAZ control the vasculature remain unclear. This study reveals that the focal adhesion protein deleted-in-liver-cancer 1 (DLC1) is a direct transcriptional target of the activated YAP/TAZ-TEAD complex. We find that substrate stiffening and VEGF stimuli promote expression of DLC1 in endothelial cells. In turn, DLC1 expression levels are YAP and TAZ dependent, and constitutive activation of YAP is sufficient to drive DLC1 expression. DLC1 is needed to limit F-actin fiber formation, integrin-based focal adhesion lifetime and integrin-mediated traction forces. Depletion of endothelial DLC1 strongly perturbs cell polarization in directed collective migration and inhibits the formation of angiogenic sprouts. Importantly, ectopic expression of DLC1 is sufficient to restore migration and angiogenic sprouting in YAP-depleted cells. Together, these findings point towards a crucial and prominent role for DLC1 in YAP/TAZ-driven endothelial adhesion remodeling and collective migration during angiogenesis.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-WWTR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Supelco
Sodium Cyclamate, analytical standard
Sigma-Aldrich
(±)-Quinuclidinyl benzilate, powder