Merck
  • Alterations of functional circuitry in aging brain and the impact of mutated APP expression.
Alterations of functional circuitry in aging brain and the impact of mutated APP expression.
Neurobiology of aging (2018-07-29)
Elaine L Bearer, Brett C Manifold-Wheeler, Christopher S Medina, Aaron G Gonzales, Frances L Chaves, Russell E Jacobs
ABSTRACT
Alzheimer's disease (AD) is a disease of aging that results in cognitive impairment, dementia, and death. Pathognomonic features of AD are amyloid plaques composed of proteolytic fragments of the amyloid precursor protein (APP) and neurofibrillary tangles composed of hyperphosphorylated tau protein. One type of familial AD occurs when mutant forms of APP are inherited. Both APP and tau are components of the microtubule-based axonal transport system, which prompts the hypothesis that axonal transport is disrupted in AD, and that such disruption impacts cognitive function. Transgenic mice expressing mutated forms of APP provide preclinical experimental systems to study AD. Here, we perform manganese-enhanced magnetic resonance imaging to study transport from hippocampus to forebrain in four cohorts of living mice: young and old wild-type and transgenic mice expressing a mutant APP with both Swedish and Indiana mutations (APPSwInd). We find that transport is decreased in normal aging and further altered in aged APPSwInd plaque-bearing mice. These findings support the hypothesis that transport deficits are a component of AD pathology and thus may contribute to cognitive deficits.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Anti-Superoxide Dismutase 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-amyloid beta peptide (MOAB-2), pan Antibody, clone 6C3, clone 6C3, from mouse
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse