Merck
  • Toxicological effects of NCKU-21, a phenanthrene derivative, on cell growth and migration of A549 and CL1-5 human lung adenocarcinoma cells.
Toxicological effects of NCKU-21, a phenanthrene derivative, on cell growth and migration of A549 and CL1-5 human lung adenocarcinoma cells.
PloS one (2017-09-26)
Hsien-Feng Liao, Chun-Hsu Pan, Pei-Yu Chou, Yi-Fong Chen, Tian-Shung Wu, Ming-Jyh Sheu, Chieh-Hsi Wu
ABSTRACT
Chemotherapy insensitivity continues to pose significant challenges for treating non-small cell lung cancer (NSCLC). The purposes of this study were to investigate whether 3,6-dimethoxy-1,4,5,8-phenanthrenetetraone (NCKU-21) has potential activity to induce effective toxicological effects in different ethnic NSCLC cell lines, A549 and CL1-5 cells, and to examine its anticancer mechanisms. Mitochondrial metabolic activity and the cell-cycle distribution were analyzed using an MTT assay and flow cytometry in NCKU-21-treated cells. NCKU-21-induced cell apoptosis was verified by Annexin V-FITC/propidium iodide (PI) double-staining and measurement of caspase-3 activity. Western blotting and wound-healing assays were applied to respectively evaluate regulation of signaling pathways and cell migration by NCKU-21. Molecular interactions between target proteins and NCKU-21 were predicted and performed by molecular docking. A colorimetric screening assay kit was used to evaluate potential regulation of matrix metalloproteinase-9 (MMP-9) activity by NCKU-21. Results indicated that NCKU-21 markedly induced cytotoxic effects that reduced cell viability via cell apoptosis in tested NSCLC cells. Activation of AMP-activated protein kinase (AMPK) and p53 protein expression also increased in both NSCLC cell lines stimulated with NCKU-21. However, repression of PI3K-AKT activation by NCKU-21 was found in CL1-5 cells but not in A549 cells. In addition, increases in phosphatidylserine externalization and caspase-3 activity also confirmed the apoptotic effect of NCKU-21 in both NSCLC cell lines. Moreover, cell migration and translational levels of the gelatinases, MMP-2 and MMP-9, were obviously reduced in both NSCLC cell lines after incubation with NCKU-21. Experimental data obtained from molecular docking suggested that NCKU-21 can bind to the catalytic pocket of MMP-9. However, the in vitro enzyme activity assay indicated that NCKU-21 has the potential to increase MMP-9 activity. Our results suggest that NCKU-21 can effectively reduce cell migration and induce apoptosis in A549 and CL1-5 cells, the toxicological effects of which may be partly modulated through PI3K-AKT inhibition, AMPK activation, an increase in the p53 protein, and gelatinase inhibition.
Sigma-Aldrich
Anti-MMP-9 Antibody, Catalytic domain, Chemicon®, from rabbit
Sigma-Aldrich
Anti-MMP-2 Antibody, catalytic domain, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Akt1/PKBα Antibody, Upstate®, from rabbit