Merck
  • Epac1 inhibition ameliorates pathological angiogenesis through coordinated activation of Notch and suppression of VEGF signaling.
Epac1 inhibition ameliorates pathological angiogenesis through coordinated activation of Notch and suppression of VEGF signaling.
Science advances (2020-01-09)
Hua Liu, Fang C Mei, Wenli Yang, Hui Wang, Eitan Wong, Jingjing Cai, Emma Toth, Pei Luo, Yue-Ming Li, Wenbo Zhang, Xiaodong Cheng
ABSTRACT
In this study, we investigated the roles of Epac1 in pathological angiogenesis and its potential as a novel therapeutic target for the treatment of vasoproliferative diseases. Genetic deletion of Epac1 ameliorated pathological angiogenesis in mouse models of oxygen-induced retinopathy (OIR) and carotid artery ligation. Moreover, genetic deletion or pharmacological inhibition of Epac1 suppressed microvessel sprouting from ex vivo aortic ring explants. Mechanistic studies revealed that Epac1 acted as a previously unidentified inhibitor of the γ-secretase/Notch signaling pathway via interacting with γ-secretase and regulating its intracellular trafficking while enhancing vascular endothelial growth factor signaling to promote pathological angiogenesis. Pharmacological administration of an Epac-specific inhibitor suppressed OIR-induced neovascularization in wild-type mice, recapitulating the phenotype of genetic Epac1 knockout. Our results demonstrate that Epac1 signaling is critical for the progression of pathological angiogenesis but not for physiological angiogenesis and that the newly developed Epac-specific inhibitors are effective in combating proliferative retinopathy.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Cytochrome P450 human, 1B1 Isozyme Microsomes, with P450 Reductase, recombinant, expressed in baculovirus infected insect cells (BTI-TN-5B1-4)
Sigma-Aldrich
MISSION® esiRNA, targeting human RAPGEF3
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), FITC conjugate, lyophilized powder
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture