Merck
  • The molecular chaperone Hsp90 maintains Golgi organization and vesicular trafficking by regulating microtubule stability.
The molecular chaperone Hsp90 maintains Golgi organization and vesicular trafficking by regulating microtubule stability.
Journal of molecular cell biology (2019-09-29)
Yuan Wu, Yubo Ding, Xiudan Zheng, Kan Liao
ABSTRACT
Hsp90 is an abundant and special molecular chaperone considered to be the regulator of many transcription factors and signaling kinases. Its high abundance is indicative of its involvement in some more fundamental processes. In this study, we provide evidence that Hsp90 is required for microtubule stabilization, Golgi organization, and vesicular trafficking. We showed that Hsp90 is bound to microtubule-associated protein 4 (MAP4), which is essential for maintaining microtubule acetylation and stabilization. Hsp90 depletion led to the decrease in MAP4, causing microtubule deacetylation and destabilization. Furthermore, in Hsp90-depleted cells, the Golgi apparatus was fragmented and anterograde vesicle trafficking was impaired, with phenotypes similar to those induced by silencing MAP4. These disruptive effects of Hsp90 depletion could be rescued by the expression of exogenous MAP4 or the treatment of trichostatin A that increases microtubule acetylation as well as stability. Thus, microtubule stability is an essential cellular event regulated by Hsp90.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Tubulin Antibody, clone YL1/2, clone YL1/2, Chemicon®, from rat
Sigma-Aldrich
Anti-MAP1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution