Merck
  • PLK1 phosphorylation of pericentrin initiates centrosome maturation at the onset of mitosis.
PLK1 phosphorylation of pericentrin initiates centrosome maturation at the onset of mitosis.
The Journal of cell biology (2011-12-21)
Kwanwoo Lee, Kunsoo Rhee
ABSTRACT
The microtubule-organizing activity of the centrosome oscillates during the cell cycle, reaching its highest level at mitosis. At the onset of mitosis, the centrosome undergoes maturation, which is characterized by a drastic expansion of the pericentriolar matrix (PCM) and a robust increase in microtubule-organizing activity. It is known that PLK1 is critical for the initiation of centrosome maturation. In this paper, we report that pericentrin (PCNT), a PCM protein, was specifically phosphorylated by PLK1 during mitosis. Phosphoresistant point mutants of PCNT did not recruit centrosomal proteins, such as CEP192, GCP-WD (γ-complex protein with WD repeats), γ-tubulin, Aurora A, and PLK1, into the centrosome during mitosis. However, centrosomal recruitment of CEP215 depended on PCNT irrespective of its phosphorylation status. Furthermore, ectopic expression of PLK1-PCNT fusion proteins induced the centrosomal accumulation of CEP192, GCP-WD, and γ-tubulin even in interphase cells, mimicking centrosome maturation. Based on these results, we propose that PLK1-mediated phosphorylation of PCNT initiates centrosome maturation by organizing the spindle pole-specific PCM lattice.
Sigma-Aldrich
Protein A, Peroxidase Conjugate, This Protein A, Peroxidase Conjugate is validated for use in Direct Immunoassay.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)