Merck
  • Anti-malarial agent artesunate inhibits TNF-alpha-induced production of proinflammatory cytokines via inhibition of NF-kappaB and PI3 kinase/Akt signal pathway in human rheumatoid arthritis fibroblast-like synoviocytes.
Anti-malarial agent artesunate inhibits TNF-alpha-induced production of proinflammatory cytokines via inhibition of NF-kappaB and PI3 kinase/Akt signal pathway in human rheumatoid arthritis fibroblast-like synoviocytes.
Rheumatology (Oxford, England) (2007-02-23)
H Xu, Y He, X Yang, L Liang, Z Zhan, Y Ye, X Yang, F Lian, L Sun
ABSTRACT
Recent studies indicate that the anti-malarial agent artemisinin and its derivatives may exert an anti-inflammatory effect. In this study, we explored the effect of artesunate, an artemisinin derivative, on tumour necrosis factor (TNF)-alpha-induced production of interleukins, IL-1beta, IL-6 and IL-8, in human rheumatoid arthritis (RA) fibroblast-like synoviocytes (FLS), and further investigated the signal mechanism by which this compound modulates those cytokines' production. RA FLS obtained from patients with active RA were stimulated with TNF-alpha and incubated with artesunate, and IL-1beta, IL-6 and IL-8 production was measured by ELISA. DNA-binding activity and nuclear translocation of nuclear factor kappa B (NF-kappaB) were measured by a sensitive multi-well colourimetric assay and confocal fluorescence microscopy, respectively. Signal transduction proteins expression was measured by western blot. Artesunate decreased the secretion of IL-1beta, IL-6 and IL-8 from TNF-alpha-stimulated RA FLS in a dose-dependent manner. Artesunate also prevented TNF-alpha-induced nuclear NF-kappaB translocation, DNA-binding activity and gene transcriptional activity, as well as phosphorylation and degradation of IkappaBalpha, but phosphorylation of p38 mitogen-activated protein kinase, extracellular signal-regulated kinase and c-Jun N-terminal kinase were unaffected. The production of IL-1beta, IL-6 and IL-8 induced by TNF-alpha was decreased by pyrrolidine dithiocarbamate (PDTC), a chemical inhibitor of NF-kappaB. These observations suggest that artesunate inhibits production of IL-1beta, IL-6 and IL-8 through inhibition of NF-kappaB signalling pathway. We also showed that artesunate prevented Akt phosphorylation. TNF-alpha-induced production of IL-1beta, IL-6 and IL-8 was hampered by treatment with the phosphatidylinositol 3 (PI3) kinase inhibitor LY294002, suggesting that inhibition of Akt activation might inhibit IL-1beta, IL-6 and IL-8 production induced by TNF-alpha. Our results indicate that artesunate exerts an anti-inflammatory effect in RA FLS and provide the evidence that artesunate may have therapeutic potential for RA.
Sigma-Aldrich
Artesunate, from Artemisia annua
Pricing and availability is not currently available.