Merck
  • Homozygous loss-of-function mutations in MNS1 cause laterality defects and likely male infertility.
Homozygous loss-of-function mutations in MNS1 cause laterality defects and likely male infertility.
PLoS genetics (2018-08-28)
Asaf Ta-Shma, Rim Hjeij, Zeev Perles, Gerard W Dougherty, Ibrahim Abu Zahira, Stef J F Letteboer, Dinu Antony, Alaa Darwish, Dorus A Mans, Sabrina Spittler, Christine Edelbusch, Sandra Cindrić, Tabea Nöthe-Menchen, Heike Olbrich, Friederike Stuhlmann, Isabella Aprea, Petra Pennekamp, Niki T Loges, Oded Breuer, Avraham Shaag, Azaria J J T Rein, Elif Yilmaz Gulec, Alper Gezdirici, Revital Abitbul, Nael Elias, Israel Amirav, Miriam Schmidts, Ronald Roepman, Orly Elpeleg, Heymut Omran
ABSTRACT
The clinical spectrum of ciliopathies affecting motile cilia spans impaired mucociliary clearance in the respiratory system, laterality defects including heart malformations, infertility and hydrocephalus. Using linkage analysis and whole exome sequencing, we identified two recessive loss-of-function MNS1 mutations in five individuals from four consanguineous families: 1) a homozygous nonsense mutation p.Arg242* in four males with laterality defects and infertility and 2) a homozygous nonsense mutation p.Gln203* in one female with laterality defects and recurrent respiratory infections additionally carrying homozygous mutations in DNAH5. Consistent with the laterality defects observed in these individuals, we found Mns1 to be expressed in mouse embryonic ventral node. Immunofluorescence analysis further revealed that MNS1 localizes to the axonemes of respiratory cilia as well as sperm flagella in human. In-depth ultrastructural analyses confirmed a subtle outer dynein arm (ODA) defect in the axonemes of respiratory epithelial cells resembling findings reported in Mns1-deficient mice. Ultrastructural analyses in the female carrying combined mutations in MNS1 and DNAH5 indicated a role for MNS1 in the process of ODA docking (ODA-DC) in the distal respiratory axonemes. Furthermore, co-immunoprecipitation and yeast two hybrid analyses demonstrated that MNS1 dimerizes and interacts with the ODA docking complex component CCDC114. Overall, we demonstrate that MNS1 deficiency in humans causes laterality defects (situs inversus) and likely male infertility and that MNS1 plays a role in the ODA-DC assembly.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammalian cells.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MNS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution