Merck
  • Phenolic 1,3-diketones attenuate lipopolysaccharide-induced inflammatory response by an alternative magnesium-mediated mechanism.
Phenolic 1,3-diketones attenuate lipopolysaccharide-induced inflammatory response by an alternative magnesium-mediated mechanism.
British journal of pharmacology (2017-02-16)
Morena Zusso, Giulia Mercanti, Federica Belluti, Rita Maria Concetta Di Martino, Andrea Pagetta, Carla Marinelli, Paola Brun, Eugenio Ragazzi, Rita Lo, Stefano Stifani, Pietro Giusti, Stefano Moro
ABSTRACT
Toll-like receptor 4 (TLR4) plays a key role in the induction of inflammatory responses both in peripheral organs and the CNS. Curcumin exerts anti-inflammatory functions by interfering with LPS-induced dimerization of TLR4-myeloid differentiation protein-2 (MD-2) complex and suppressing pro-inflammatory mediator release. However, the inhibitory mechanism of curcumin remains to be defined. Binding of bis-demethoxycurcumin (GG6) and its cyclized pyrazole analogue (GG9), lacking the 1,3-dicarbonyl function, to TLR4-MD-2 was determined using molecular docking simulations. The effects of these compounds on cytokine release and NF-κB activation were examined by ELISA and fluorescence staining in LPS-stimulated primary microglia. Interference with TLR4 dimerization was assessed by immunoprecipitation in Ba/F3 cells. Both curcumin analogues bound to the hydrophobic region of the MD-2 pocket. However, only curcumin and GG6, both possessing the 1,3-diketone moiety, inhibited LPS-induced TLR4 dimerization, activation of NF-κB and secretion of pro-inflammatory cytokines in primary microglia. Consistent with the ability of 1,3-diketones to coordinate divalent metal ions, LPS stimulation in a low magnesium environment decreased pro-inflammatory cytokine release and NF-κB p65 nuclear translocation in microglia and decreased TLR4-MD-2 dimerization in Ba/F3 cells. Curcumin and GG6 also significantly reduced cytokine output in contrast to the pyrazole analogue GG9. These results indicate that phenolic 1,3-diketones, with a structural motif able to coordinate magnesium ions, can modulate LPS-mediated TLR4-MD-2 signalling. Taken together, these studies identify a previously uncharacterized mechanism involving magnesium, underlying the inflammatory responses to LPS.
Sigma-Aldrich
Anti-GFP (Green Fluorescent Protein) Antibody, Upstate®, from chicken
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)