
Vitamin D2 (6,19,19-d3) solution

Synonym(s):
Calciferol (6,19,19-d3) solution, Ercalciol (6,19,19-d3) solution, Ergocalciferol (6,19,19-d3) solution
Empirical Formula (Hill Notation):
C28D3H41O
Molecular Weight:
399.67
EC Number:
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-2 of 2