
Tyrosol

Synonym(s):
2-(4-Hydroxyphenyl)ethanol, 4-Hydroxyphenethyl alcohol, Tyrosol, p-HPEA
Linear Formula:
HOC6H4CH2CH2OH
CAS Number:
Molecular Weight:
138.16
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing