![TSKgel® SWXL Top-Off Gel phase diol, 5 μm particle size, bottle of 1 g](/deepweb/assets/sigmaaldrich/product/images/513/973/52808a38-c28f-4c0a-b317-dc365391424f/640/52808a38-c28f-4c0a-b317-dc365391424f.jpg)
TSKgel® SWXL (5 µm) HPLC Columns
![TSKgel® SWXL Top-Off Gel phase diol, 5 μm particle size, bottle of 1 g](/deepweb/assets/sigmaaldrich/product/images/513/973/52808a38-c28f-4c0a-b317-dc365391424f/640/52808a38-c28f-4c0a-b317-dc365391424f.jpg)
Synonym(s):
TSKgel® SWXL Columns
Products
Product Category
Manufacturer
Physical Form
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-3 of 3