Tryptic Soy Agar - Ready-to-use Contact Plates
Products
Product Category
Brand
Color
Physical Form
Agency
Chemical Composition
Diameter (mm)
Quality Segment
Application
Plate Material
Microorganism Suitability
Package Size
Sterility
Sterilization Method
Showing 1-10 of 10
Compare
Product Number
Description
Pricing