
Tris acidimetric

Synonym(s):
Trizma® base, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol
Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
pH Value
Physical Form
pK Value
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing