
Tigogenin

Synonym(s):
(25R)-5α-Spirostan-3β-ol
Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing