
Thiamine phosphoric acid ester chloride dihydrate

Synonym(s):
Thiamine monophosphate chloride dihydrate, Aneurine monophosphate chloride
Empirical Formula (Hill Notation):
C12H18ClN4O4PS · 2H2O
CAS Number:
Molecular Weight:
416.82
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing