![Tetra-n-butylammonium hydrogen sulfate for synthesis](/deepweb/assets/sigmaaldrich/product/images/148/626/310ea303-a636-4c39-bff5-8ec913b09849/640/310ea303-a636-4c39-bff5-8ec913b09849.jpg)
Tetra-n-butylammonium hydrogen sulfate
![Tetra-n-butylammonium hydrogen sulfate for synthesis](/deepweb/assets/sigmaaldrich/product/images/148/626/310ea303-a636-4c39-bff5-8ec913b09849/640/310ea303-a636-4c39-bff5-8ec913b09849.jpg)
Synonym(s):
Tetra-n-butylammonium hydrogen sulfate
Empirical Formula (Hill Notation):
C16H37NO4S
CAS Number:
Molecular Weight:
339.53
Products
Brand
Formula Weight
pH Value
Physical Form
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing