![Tetramethrin mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/385/685/bec42ca6-5ae1-4a09-b089-7cf0183d3fa4/640/bec42ca6-5ae1-4a09-b089-7cf0183d3fa4.png)
Tetramethrin
![Tetramethrin mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/385/685/bec42ca6-5ae1-4a09-b089-7cf0183d3fa4/640/bec42ca6-5ae1-4a09-b089-7cf0183d3fa4.png)
Empirical Formula (Hill Notation):
C19H25NO4
CAS Number:
Molecular Weight:
331.41
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2