Skip to Content
Merck
Tetraethylammonium bromide suitable for ion pair chromatography, LiChropur™, ≥99.0%

Tetraethylammonium bromide

Tetraethylammonium bromide suitable for ion pair chromatography, LiChropur™, ≥99.0%
Synonym(s):
TEA bromide, Tetraethylammonium bromide
Linear Formula:
(C2H5)4N(Br)
CAS Number:
Molecular Weight:
210.16
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Technique