
Testosterone isocaproate

Synonym(s):
4-Androsten-17β-ol-3-one 17-(4-methylpentanoate), Testosterone 4-methylvalerate
Empirical Formula (Hill Notation):
C25H38O3
CAS Number:
Molecular Weight:
386.57
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing