Skip to Content
Merck
Tannic acid SAJ first grade

Tannic acid

Tannic acid SAJ first grade
Synonym(s):
Gallotannin, Tannin
Empirical Formula (Hill Notation):
C76H52O46
CAS Number:
Molecular Weight:
1701.20

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade