Skip to Content
Merck
Sodium diethyldithiocarbamate trihydrate PESTANAL®, analytical standard

Sodium diethyldithiocarbamate trihydrate

Sodium diethyldithiocarbamate trihydrate PESTANAL®, analytical standard
Synonym(s):
Cupral, Diethyldithiocarbamic acid sodium salt, Sodium diethyldithiocarbamate trihydrate
Linear Formula:
(C2H5)2NCSSNa · 3H2O
CAS Number:
Molecular Weight:
225.31
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique